Learn More
Biomatik Corporation Dog Vascular Cell Adhesion Molecule 1 (VCAM1) ELISA Kit
Supplier:  Biomatik Corporation EKU08513
Encompass_Preferred
Dog Vascular Cell Adhesion Molecule 1 (VCAM1) ELISA Kit for detecting VCAM1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Dog (Canis familiaris; Canine). Detection Range: 3.12-200ng/ml. Sensitivity: 1.25ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q28260. Gene ID: 403982. Target Synonyms: CD106; INCAM-100; L1CAM. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5519