Learn More
American Research Products Inc Dog Vascular Endothelial cell
Catalog No. 5000329020
Encompass
Dog Vascular Endothelial cell Growth Factor VEGF ELISA KIT from CUSABIO Detection Range 3 12 pg/mL-200 pg/mL Sensitivity 0 78 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29020 Supplier American Research Products Inc Supplier No. CSBE11744C
Dog Vascular Endothelial cell Growth Factor VEGF ELISA KIT from CUSABIO Detection Range 3 12 pg/mL-200 pg/mL Sensitivity 0 78 pg/mL Sample serum plasma tissue homogenates Method Sandwich