Learn More
Biomatik Corporation Dog Vascular Endothelial Growth Factor D (VEGFD) ELISA Kit
Supplier:  Biomatik Corporation EKU11870
Encompass_Preferred
Dog Vascular Endothelial Growth Factor D (VEGFD) ELISA Kit for detecting VEGFD in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Dog (Canis familiaris; Canine). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.0pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: A0A8C0YXI0. Gene ID: 491749. Target Synonyms: FIGF; VEGF-D; C-Fos Induced Growth Factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7784