Learn More
Diagnostic Biosystems DOG11 DOG 1
Catalog No. 502407607
Encompass_Preferred
DOG1 is expressed ubiquitously in gastrointestinal stromal tumors irrespective of c-kit or PDGFR alpha mutation status. Reactivity for DOG1 may aid in the diagnosis of gastrointestinal stromal tumors, including PDGFRA mutants that fail to express c-kit antigen, and lead to appropriate treatment with imatinib mesylate, an inhibitor of the c-kit tyrosine kinase.
Catalog No. 50-240-7607 Supplier Diagnostic Biosystems Supplier No. PDM170
DOG1 is expressed ubiquitously in gastrointestinal stromal tumors irrespective of c-kit or PDGFR alpha mutation status. Reactivity for DOG1 may aid in the diagnosis of gastrointestinal stromal tumors, including PDGFRA mutants that fail to express c-kit antigen, and lead to appropriate treatment with imatinib mesylate, an inhibitor of the c-kit tyrosine kinase.