Learn More
Avanti Polar Lipids DOGS-NTA IN CHLOROFORM, 10MGD
Catalog No. NC9735063
Encompass
1,2-dioleoyl-sn-glycero-3-[(N-(5-amino-1-carboxypentyl)iminodiacetic acid)succinyl] (nickel salt). 231615-77-3
Catalog No. NC9735063 Supplier Avanti Polar Lipids Supplier No. 790404C10MG
1,2-dioleoyl-sn-glycero-3-[(N-(5-amino-1-carboxypentyl)iminodiacetic acid)succinyl] (nickel salt). 231615-77-3