Learn More
Aviva Systems Biology Corp DOHH Antibody - N-terminal region (ARP60855_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60855_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DOHH. It was validated on Western Blot. . Alternate names: HLRC1, MGC4293, hDOHHApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat, Yeast, Zebrafish
Catalog No. 50-287-190