Learn More
Dojindo Molecular Technologies Inc Dojindo Labeling Kit, HiLyte Fluor™ 647, NH2, 3 Samples
SDP
Catalog No. NC3985568
Encompass
This labeling kit is primarily utilized for preparing red fluorescence-labeled proteins, such as IgG for immunostaining, and cellular proteins for tracing. It includes NH2-reactive HiLyte Fluor™ 647, which has a succinimidyl group (NHS) that reacts with amino groups on proteins or other molecules. This kit contains all necessary reagents for labeling.
  • Only 1 hour to recover conjugates
  • All processes are conducted in a single filtration tube
  • High recovery of conjugates
  • Applicable for 50 to 200 μg IgG
  • Simple labeling process, forming covalent bond with target protein without activation
  • Filtration tube removes small molecules that interfere with the assay or labeling reaction
  • Comprehensive kit contains all necessary reagents for labeling, including storage buffer for conjugates
Catalog No. NC3985568 Supplier Dojindo Molecular Technologies Inc Supplier No. LK1510
This labeling kit is primarily utilized for preparing red fluorescence-labeled proteins, such as IgG for immunostaining, and cellular proteins for tracing. It includes NH2-reactive HiLyte Fluor™ 647, which has a succinimidyl group (NHS) that reacts with amino groups on proteins or other molecules. This kit contains all necessary reagents for labeling.
  • Only 1 hour to recover conjugates
  • All processes are conducted in a single filtration tube
  • High recovery of conjugates
  • Applicable for 50 to 200 μg IgG
  • Simple labeling process, forming covalent bond with target protein without activation
  • Filtration tube removes small molecules that interfere with the assay or labeling reaction
  • Comprehensive kit contains all necessary reagents for labeling, including storage buffer for conjugates