Learn More
Aviva Systems Biology Corp DOK4 Antibody - N-terminal region (ARP60243_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60243_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DOK4. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human DOK4. Alternate names: FLJ10488ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-984