Learn More
Aviva Systems Biology Corp DOK6 Antibody - middle region (ARP55502_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55502_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DOK6. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human DOK6. Alternate names: DOK5L, HsT3226, MGC20785ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-045