Learn More
Aviva Systems Biology Corp DOK7 Antibody - C-terminal region (ARP61801_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61801_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DOK7. It was validated on Western Blot. . Alternate names: C4orf25, CMS1B, FLJ33718, FLJ39137, FLJ90556ApplicationWB. Species reactivity: Human
Catalog No. 50-287-471