Learn More
Apexbio Technology LLC Dolasetron, 5mg CAS# 115956-12-2
SDP
Supplier:  Apexbio Technology LLC B87185
Encompass_Preferred
Dolasetron is a serotonin 5-HT3 receptor antagonist, the main role is to reduce vagal activity, used to treat nausea and vomiting after chemotherapy. The drug has no effect on dopamine receptors or muscarinic receptors. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-199-8463