Learn More
Selleck Chemical LLC Dolasetron Mesylate
SDP
Catalog No. 5000003701
Encompass_Preferred
Dolasetron Mesylate (Anzemet hydrate MDL-73147EF) is a serotonin 5-HT3 receptor antagonist used to treat nausea and vomiting following chemotherapy
Catalog No. 50-000-03701 Supplier Selleck Chemical LLC Supplier No. S56795MG
Dolasetron Mesylate (Anzemet hydrate MDL-73147EF) is a serotonin 5-HT3 receptor antagonist used to treat nausea and vomiting following chemotherapy