Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Dolichos Biflorus (Horse Gram) Agglutinin (DBA), fluorescein (FITC)

Catalog No. L32474
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L32474 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. L32474 Supplier Invitrogen™ Supplier No. L32474
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bright-green DBA-fluorophor conjugate

  • Dolichos Biflorus (Horse Gram) Agglutinin (DBA), fluorescein (FITC), is a bright-green DBA-fluorophor conjugate. DBA has been used to establish secretor status in blood-group-A individuals by hemagglutination inhibition techniques and for blood typing.
  • This lectin has also been used as a general marker of developing renal collecting ducts. It has also been used as a marker of cell surface change in murine embryonic stem cells during early differentiation.
  • Very bright FITC conjugate detected with traditional FITC/GFP filter set, Ex/Em: 495/515 nm
  • Applications: immunofluorescence, glycobiology
  • Sugar specificity: N-acetylgalactosamine
  • Inhibiting/eluting sugar: 200 mM N-acetylgalactosamine
  • Blood group specificity: A1 >>A2
  • Unconjugated lectin molecular weight: 111 KDa
  • Immunofluorescence working concentration: 5–20 μg/mL
  • DBA-FITC is provided at a 2 mg/mL concentration in 10 mM HEPES, 0.15 M NaCl, pH 7.5, 0.08% sodium azide, 0.1 mM Ca2+. If a precipitate forms upon long-term storage at 4°C, warm to 37°C and centrifuge before use.
TRUSTED_SUSTAINABILITY

Specifications

Conjugate FITC
Quantity 2 mg
Source Horse gram
Content And Storage When stored at or below 2–8°C in the dark, the product is stable for 6 months after reciept.

Shipping Condition Approved for shipment at Room Temperature or on Wet Ice
Form Lyophilized
Protein Family Lectins

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.