Learn More
Kingfisher Biotech Inc Equine CCL11 (Eotaxin-1) ELISA - Less than or equal to 20 plates (96-well)
SDP
Supplier:  Kingfisher Biotech Inc DIY1549P004
Encompass_Preferred
The Equine CCL11 (Eotaxin-1) ELISA contains capture antibody, standard, and detection antibody for development of a Equine CCL11 ELISA. The antibodies have been determined to function in an ELISA with the standard provided. Optimal buffers, concentrations, incubation times, incubation temperatures, and methods for the ELISA have not been determined. This allows for complete flexibility to provide optimal dynamic range and sensitivity based on your sample type. Alternate names include CCL11, SCYA11, C-C motif chemokine ligand 11, Eotaxin-1, eosinophil chemotactic protein. The quantities of components provided are not matched. Components may also be purchased separately. For research use only. Made in the USA
Catalog No. 50-228-5520