Learn More
Raybiotech Inc C-Series Dolphin Cytokine Array C1 Kit.
Catalog No. 501948450
Encompass_Preferred
Detects 10 Dolphin Cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: IFN-gamma, IL-1 beta, IL-1 Ra, IL-2, IL-6, IL-8, IL-13, IL-17A, IP-10, TNF alpha
Catalog No. 50-194-8450 Supplier Raybiotech Inc Supplier No. AADCYT12
Detects 10 Dolphin Cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: IFN-gamma, IL-1 beta, IL-1 Ra, IL-2, IL-6, IL-8, IL-13, IL-17A, IP-10, TNF alpha