Learn More
Raybiotech Inc G-Series Dolphin Cytokine Array 1 Kit.
Supplier:  Raybiotech Inc GSDCYT11
Encompass_Preferred
Detects 10 Dolphin Cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: IFN gamma, IL-1 beta, IL-1ra, IL-2, IL-6, IL-8, IL-13, IL-17, IP-10, TNF alpha
Catalog No. 50-195-0346