Learn More
Kingfisher Biotech Inc Bovine CD40 Ligand ELISA - Less than or equal to 10 plates (96-well)
SDP
Supplier:  Kingfisher Biotech Inc DIY1159P003
Encompass_Preferred
The Bovine CD40 Ligand ELISA contains capture antibody, standard, and detection antibody for development of a Bovine CD40 Ligand ELISA. The antibodies have been determined to function in an ELISA with the standard provided. Optimal buffers, concentrations, incubation times, incubation temperatures, and methods for the ELISA have not been determined. This allows for complete flexibility to provide optimal dynamic range and sensitivity based on your sample type. The Bovine CD40 Ligand ELISA can also be used to measure Bison and Wild Yak CD40 Ligand. Alternate names include CD40LG, CD154, CD40L, HIGM1, IGM, IMD3, T-BAM, TNFSF5, TRAP, gp39, hCD40L, CD40 ligand. The quantities of components provided are not matched. Components may also be purchased separately. For research use only. Made in the USA
Catalog No. 50-228-5345