Learn More
Cayman Chemical DolutegravIr 10mg
SDP
Catalog No. 502986341
Encompass_Preferred
A potent inhibitor of HIV integrase (IC50 = 2.7 nM for HIV-1 integrase-catalyzed strand transfer in vitro); inhibits HIV-1 viral replication (EC50 = 0.51 nM) in PMBCs; CC50 values are 189 and 52 µM in unstimulated and stimulated PBMCs; prevents replication of several HIV-1 strains (EC50s = 0.36-2.1 nM) that are resistant to NRTIs, NNRTIs, and protease inhibitors and impairs their ability to infect CIP4 cells
Catalog No. 50-298-6341 Supplier Cayman Chemical Supplier No. 2219110 MG
A potent inhibitor of HIV integrase (IC50 = 2.7 nM for HIV-1 integrase-catalyzed strand transfer in vitro); inhibits HIV-1 viral replication (EC50 = 0.51 nM) in PMBCs; CC50 values are 189 and 52 µM in unstimulated and stimulated PBMCs; prevents replication of several HIV-1 strains (EC50s = 0.36-2.1 nM) that are resistant to NRTIs, NNRTIs, and protease inhibitors and impairs their ability to infect CIP4 cells