Learn More
Selleck Chemical LLC Dolutegravir S2667-50mg
SDP
Catalog No. 503111785
Encompass_Preferred
Dolutegravir is a two-metal-binding HIV integrase inhibitor with IC50 of 2 7 nM in a cell-free assay modest activity against raltegravir-resistant signature mutants Y143R Q148K N155H and G140S/Q148H
Catalog No. 50-311-1785 Supplier Selleck Chemical LLC Supplier No. S266750MG
Dolutegravir is a two-metal-binding HIV integrase inhibitor with IC50 of 2 7 nM in a cell-free assay modest activity against raltegravir-resistant signature mutants Y143R Q148K N155H and G140S/Q148H