Learn More
Selleck Chemical LLC Dolutegravir (GSK1349572) 5mg 1051375-16-6 S/GSK1349572
SDP
Catalog No. 501362717
Encompass_Preferred
Dolutegravir (GSK1349572, S/GSK1349572) is a two-metal-binding HIV integrase inhibitor with IC50 of 2.7 nM in a cell-free assay, modest activity against raltegravir-resistant signature mutants Y143R, Q148K, N155H, and G140S/Q148H. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-2717 Supplier Selleck Chemical LLC Supplier No. S26675MG
Dolutegravir (GSK1349572, S/GSK1349572) is a two-metal-binding HIV integrase inhibitor with IC50 of 2.7 nM in a cell-free assay, modest activity against raltegravir-resistant signature mutants Y143R, Q148K, N155H, and G140S/Q148H. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.