Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dolutegravir intermediate-1 | 1335210-23-5 | MFCD28978322 | 99.8% | 315.28 | C13H17NO8 | 500 G
SDP

Catalog No. 5000215366
Encompass_Preferred

Dolutegravir intermediate-1 is a synthetic intermediate used in the research and synthesis of dolutegravir, an HIV-1 integrase inhibitor. It is supplied for research use only and includes analytical specifications suitable for laboratory development and quality control.

  • CAS number 1335210-23-5 for precise identification.
  • Purity 99.8% as supplied.
  • Molecular weight 315.28 g/mol.
  • Chemical formula C13H17NO8.
  • Appearance solid, white to light yellow.
  • Available in multiple package sizes up to 500 G.
  • Store dry and cool; in solvent store at -80°C (2 years) or -20°C (1 year).
  • Safety data sheet available for handling and hazard information.

Catalog No. 50-002-15366 Supplier Medchemexpress LLC Supplier No. HY100083500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dolutegravir intermediate-1 is a synthetic intermediate used in the research and synthesis of dolutegravir, an HIV-1 integrase inhibitor. It is supplied for research use only and includes analytical specifications suitable for laboratory development and quality control.

  • CAS number 1335210-23-5 for precise identification.
  • Purity 99.8% as supplied.
  • Molecular weight 315.28 g/mol.
  • Chemical formula C13H17NO8.
  • Appearance solid, white to light yellow.
  • Available in multiple package sizes up to 500 G.
  • Store dry and cool; in solvent store at -80°C (2 years) or -20°C (1 year).
  • Safety data sheet available for handling and hazard information.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.