Learn More
Medchemexpress LLC Dolutegravir intermediate-1 | 1335210-23-5 | MFCD28978322 | 99.8% | 315.28 g/mol | C13H17NO8 | 100 G
SDP
Catalog No. 5000215183
Encompass_Preferred
Dolutegravir intermediate-1 is a synthetic building block used in the synthesis of the HIV integrase inhibitor dolutegravir. It is supplied as a high-purity solid or as a ready-to-use solution for research and development applications, with documentation available for safe handling and quality verification.
  • High purity (99.8%) for reliable synthetic performance.
  • Available as solid and as 10 mM solution in DMSO for flexible use.
  • Multiple package sizes up to 500 g to support scale-up.
  • Documented safety and quality data including SDS and COA.
  • Suitable for research and process development workflows.
Catalog No. 50-002-15183 Supplier Medchemexpress LLC Supplier No. HY100083100G
Dolutegravir intermediate-1 is a synthetic building block used in the synthesis of the HIV integrase inhibitor dolutegravir. It is supplied as a high-purity solid or as a ready-to-use solution for research and development applications, with documentation available for safe handling and quality verification.
  • High purity (99.8%) for reliable synthetic performance.
  • Available as solid and as 10 mM solution in DMSO for flexible use.
  • Multiple package sizes up to 500 g to support scale-up.
  • Documented safety and quality data including SDS and COA.
  • Suitable for research and process development workflows.