Learn More
Aviva Systems Biology Corp DXO Antibody - N-terminal region (ARP52086_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52086_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Dom3z. It was validated on Western Blot. . Alternate names: MGC95256ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-632