Learn More
Medchemexpress LLC HY-16012 5mg Medchemexpress, Domatinostat (tosylate) CAS:1186222-89-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY160125MG
Encompass_Preferred
Medchemexpress, HY-16012 5mg Domatinostat (tosylate) CAS:1186222-89-8 Domatinostat tosylate (4SC-202) is a selective class I HDAC inhibitor with IC50 of 1.20 μM, 1.12 μM, and 0.57 μM for HDAC1, HDAC2, and HDAC3, respectively. It also displays inhibitory activity against Lysine specific demethylase 1 (LSD1). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1861