Learn More
MCR Safety Dominator™ 3
Catalog No. 19812644 Shop All MCR Safety Products
Change view
Click to view available options
Lens Coating:
Anti-fog
Scratch-resistant
Lens Tint:
Amber
Blue
Clear
Gray
Indoor/Outdoor
Mirror
Frame Color:
Black
Blue, Yellow
Clear
Gun Metal
19 product options available for selection
Product selection table with 19 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Coating Lens Tint Frame Color
19-812-644 Anti-fog Clear Gun Metal
19-812-645 Anti-fog Gray Gun Metal
19-812-647 - Mirror Gun Metal
19-812-671 - Gray Gun Metal
19-812-672 - Mirror Gun Metal
19-812-673 - Clear Clear
19-812-674 - Gray Clear
19-812-675 - Blue Clear
19-812-676 - Mirror Clear
19-812-677 - Mirror Clear
19-812-678 - Indoor/Outdoor Clear
19-812-679 - Clear Black
19-812-680 - Gray Black
19-812-682 Scratch-resistant Gray Black
19-812-683 - Gray Black
19-812-684 - Clear Blue, Yellow
19-812-685 Scratch-resistant Mirror Blue, Yellow
19-812-646 - Indoor/Outdoor Gun Metal
19-812-681 Anti-fog Amber Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 19 options available.
19 missing translation for 'options'
This item is not returnable. View return policy
Catalog No. 19-812-644 missing translation for 'mfr' MCR Safety missing translation for 'supplierNo' DM1310PF
Add to Cart
Add to Cart
This item is not returnable. View return policy
The Dominator™ series core feature includes wire core independently adjustable nose pads to fit most facial profiles. Meets or exceeds Z87+ and Military High Velocity Impact standards. Dominate your day with adjustability, protection, and comfort!
  • Italian fashion, sport focused design
  • Co-injected wire core independently adjustable nose pads
  • Co-injected soft wire core adjustable TPR temples
  • MAX3™, MAX6™, or BOSSMAN™ Mirror coatings
  • Meets or Exceeds Z87+ & Military High Velocity
  • Gun Metal/Red TPR, Clear/Blue TPR, Black/Black TPR, or Yellow/Blue TPR
Certifications
CSA
Compliance
Meets ANSI Z87.1-2015, CSA Z94.3 and MIL - PRF - 31013 standards.
Specifications
Lens Coating Anti-fog
Lens Tint Clear
For Use With (Application) Manufacturing
Frame Color Gun Metal
No. per Pack 1 Pair
Lens Material Polycarbonate
Frame Material Polycarbonate