Learn More
MCR Safety Dominator™ 3
Catalog No. 19812685 Shop All MCR Safety Products
Encompass_Preferred
Change view
Click to view available options
Lens Coating:
Anti-fog
Scratch-resistant
Lens Tint:
Blue
Clear
Gray
Indoor/Outdoor
Mirror
Frame Color:
Black
Blue, Yellow
Clear
Gun Metal
17 product options available for selection
Product selection table with 17 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Coating Lens Tint Frame Color
19-812-685 Scratch-resistant Mirror Blue, Yellow
19-812-644 Anti-fog Clear Gun Metal
19-812-645 Anti-fog Gray Gun Metal
19-812-647 Scratch-resistant Mirror Gun Metal
19-812-671 Scratch-resistant Gray Gun Metal
19-812-672 Scratch-resistant Mirror Gun Metal
19-812-673 Anti-fog Clear Clear
19-812-674 Anti-fog Gray Clear
19-812-675 Anti-fog Blue Clear
19-812-676 Scratch-resistant Mirror Clear
19-812-677 Scratch-resistant Mirror Clear
19-812-678 Scratch-resistant Indoor/Outdoor Clear
19-812-679 Anti-fog Clear Black
19-812-680 Anti-fog Gray Black
19-812-683 Scratch-resistant Gray Black
19-812-684 Anti-fog Clear Blue, Yellow
19-812-682 Scratch-resistant Gray Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 17 options available.
17 options
This item is not returnable. View return policy
Catalog No. 19-812-685 Supplier MCR Safety Supplier No. DM1348B
This item is not returnable. View return policy
The Dominator™ series core feature includes wire core independently adjustable nose pads to fit most facial profiles. Meets or exceeds Z87+ and Military High Velocity Impact standards. Dominate your day with adjustability, protection, and comfort!
  • Italian fashion, sport focused design
  • Co-injected wire core independently adjustable nose pads
  • Co-injected soft wire core adjustable TPR temples
  • MAX3™, MAX6™, or BOSSMAN™ Mirror coatings
  • Meets or Exceeds Z87+ & Military High Velocity
  • Gun Metal/Red TPR, Clear/Blue TPR, Black/Black TPR, or Yellow/Blue TPR
Certifications
CSA
Compliance
Meets ANSI Z87.1-2015, CSA Z94.3 and MIL - PRF - 31013 standards.
Specifications
Lens Coating Scratch-resistant
Lens Tint Mirror
Product Type Safety Glasses
For Use With (Application) Manufacturing
Frame Color Blue, Yellow
No. per Pack 1 Pair
Material Polycarbonate, TPR
Disposable Reusable
Lens Material Polycarbonate
Frame Material Polycarbonate
UV Protection 100% UV
Frame Type Wrap-around