Learn More
MCR Safety™ Dominator DM3-MD Clear UV-AF Lens
Catalog No. 19034399 Shop All MCR Safety Products
Encompass_Preferred
Change view
Click to view available options
Lens Coating:
Anti-fog, Impact-resistant
Catalog No. Lens Coating
19-034-399 Anti-fog, Impact-resistant
1 options
Catalog No. 19-034-399 Supplier MCR Safety™ Supplier No. DM3MD10AF
Dominator™ DM3-MD series - Metal detectable / X-Ray detectable, clear UV-AF™ anti-fog lens, clear frame with blue temples, earplug retaining technology.
  • Patent pending design
  • Metal detectable safety glass built and tested for the food industry
  • Metal detectable steel strip embedded in lens
  • Metal detectable steel temple screws
  • Metal detectable, co-injected steel wire core independently adjustable temples
  • X-ray detectable
  • Metal in all frame components
  • ERT Earplug Retaining Technolgy holds earplugs in place
  • Available in Duramass™ scratch resistant or UV-AF™ anti-fog coatings
  • No removable components
  • Rugged polycarbonate lenses filter 99.9% of harmful U.V. rays
  • CAN/CSA Z94.3 compliant
  • Meets ANSI/ISEA Z87.1 2020 standard
Specifications
Lens Coating Anti-fog, Impact-resistant
Lens Tint Clear
Product Type Glasses
Material Polycarbonate
Disposable Reusable
Lens Material Plastic
Frame Material Polycarbonate
UV Protection U6
Frame Type Frameless
Autoclavable Not Autoclavable