Learn More
DOMINO AMJET INC DOMINO AMJET INK IC-2CL158 EA
Catalog No. NC1827040
Encompass
  • Clear UV-fluorescent ink for high visibility.
  • Retort resistant for durability in industrial applications.
  • Designed for continuous inkjet printing technology.
  • Part of the Ax-Series technology for optimal compatibility.
  • Solvent based formulation for superior performance.
  • Net weight: 0.781 kg.
  • Volume: 0.825 L.
  • Category: Consumable product.
  • Sold as: 1 each for convenience.
Catalog No. NC1827040 Supplier DOMINO AMJET INC Supplier No. IC2CL158
  • Clear UV-fluorescent ink for high visibility.
  • Retort resistant for durability in industrial applications.
  • Designed for continuous inkjet printing technology.
  • Part of the Ax-Series technology for optimal compatibility.
  • Solvent based formulation for superior performance.
  • Net weight: 0.781 kg.
  • Volume: 0.825 L.
  • Category: Consumable product.
  • Sold as: 1 each for convenience.