Learn More
Medchemexpress LLC Domiphen bromide | 538-71-6 | 98.7% | 1 ML
SDP
Catalog No. 5000411389
Encompass_Preferred
Domiphen bromide is a cationic active quaternary ammonium salt that inhibits HERG channels, aminopeptidase-like enzymes, Escherichia coli alkaline phosphatase, and α-chymotrypsin. It exhibits antibacterial, antimalarial, and disinfectant properties, and acts as a synergist for Colistin. This compound can be utilized as a chemical preservative and cationic surfactant, and is also relevant for research into bacterial infectious diseases such as pharyngitis, thrush, and oral ulcers.
  • Cationic active quaternary ammonium salt
  • Inhibitor of HERG channels (IC50: 9 nM), aminopeptidase-like enzymes, Escherichia coli alkaline phosphatase, and α-chymotrypsin
  • Antibacterial, antimalarial, and disinfectant properties
  • Synergist of colistin
  • Can be used as a chemical preservative and cationic surfactant
  • Applicable in research of bacterial infectious diseases like pharyngitis, thrush, and oral ulcers
Catalog No. 50-004-11389 Supplier Medchemexpress LLC Supplier No. HYB146710MM/1ML
Domiphen bromide is a cationic active quaternary ammonium salt that inhibits HERG channels, aminopeptidase-like enzymes, Escherichia coli alkaline phosphatase, and α-chymotrypsin. It exhibits antibacterial, antimalarial, and disinfectant properties, and acts as a synergist for Colistin. This compound can be utilized as a chemical preservative and cationic surfactant, and is also relevant for research into bacterial infectious diseases such as pharyngitis, thrush, and oral ulcers.
  • Cationic active quaternary ammonium salt
  • Inhibitor of HERG channels (IC50: 9 nM), aminopeptidase-like enzymes, Escherichia coli alkaline phosphatase, and α-chymotrypsin
  • Antibacterial, antimalarial, and disinfectant properties
  • Synergist of colistin
  • Can be used as a chemical preservative and cationic surfactant
  • Applicable in research of bacterial infectious diseases like pharyngitis, thrush, and oral ulcers