Learn More
Selleck Chemical LLC Domiphen Bromide S4186-1g
SDP
Catalog No. 503108578
Encompass_Preferred
Domiphen bromide (NSC-39415) is a quaternary ammonium antiseptic with actions as a cationic surfactant
Catalog No. 50-310-8578 Supplier Selleck Chemical LLC Supplier No. S41861G
Domiphen bromide (NSC-39415) is a quaternary ammonium antiseptic with actions as a cationic surfactant