Learn More
Cayman Chemical Domperidone, Purity: ≥98%, CAS: 57808-66-9, 100 mg
SDP
Catalog No. 502985179
Encompass_Preferred
A dopamine D2 receptor antagonist (Ki = 0.3 nM in CHO cells expressing the rat receptor); selective for dopamine D2 over D3 receptors (Ki = 9.5 nM); inhibits dipropyl dopamine-induced femoral vasodilation in dogs at 0.5-5 μg/kg; prevents dopamine-induced decreases in pentagastrin-induced gastric antral motility in dogs at 0.5 mg/kg; inhibits apomorphine-induced emesis in dogs (ED50 = 0.031 mg/kg, p.o.); increases serum levels of prolactin in male rats
Catalog No. 50-298-5179 Supplier Cayman Chemical Supplier No. 18875100 MG
A dopamine D2 receptor antagonist (Ki = 0.3 nM in CHO cells expressing the rat receptor); selective for dopamine D2 over D3 receptors (Ki = 9.5 nM); inhibits dipropyl dopamine-induced femoral vasodilation in dogs at 0.5-5 μg/kg; prevents dopamine-induced decreases in pentagastrin-induced gastric antral motility in dogs at 0.5 mg/kg; inhibits apomorphine-induced emesis in dogs (ED50 = 0.031 mg/kg, p.o.); increases serum levels of prolactin in male rats