Learn More
Medchemexpress LLC HY-B0411 100mg Medchemexpress, Domperidone CAS:57808-66-9 Purity:>98%
SDP
Catalog No. 502082525
Encompass_Preferred
Medchemexpress, HY-B0411 100mg Domperidone CAS:57808-66-9 Domperidone (R33812) is a selective dopamine-2 receptor antagonist. Domperidone acts as an antiemetic and a prokinetic agent through its effects on the chemoreceptor trigger zone and motor function of the stomach and small intestine. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2525 Supplier Medchemexpress LLC Supplier No. HYB0411100MG
Medchemexpress, HY-B0411 100mg Domperidone CAS:57808-66-9 Domperidone (R33812) is a selective dopamine-2 receptor antagonist. Domperidone acts as an antiemetic and a prokinetic agent through its effects on the chemoreceptor trigger zone and motor function of the stomach and small intestine. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.