Learn More
Apexbio Technology LLC Domperidone 57808-66-9 100mg
SDP
Catalog No. 502240809
Encompass_Preferred
Domperidone is a small-molecule antagonist targeting the dopamine D2 receptor It is designed to inhibit peripheral dopamine receptors thereby enhancing gastrointestinal motility and accelerating gastric emptying Domperidone exerts its biological activity primarily through peripheral D2 receptor antagonism Based on these pharmacological properties domperidone holds research potential in the investigation and management of gastroparesis and as a comparative compound for mechanistic studies of gastric motility disorders
This item is not returnable. View return policy
Catalog No. 50-224-0809 Supplier Apexbio Technology LLC Supplier No. B1481100
Domperidone is a small-molecule antagonist targeting the dopamine D2 receptor It is designed to inhibit peripheral dopamine receptors thereby enhancing gastrointestinal motility and accelerating gastric emptying Domperidone exerts its biological activity primarily through peripheral D2 receptor antagonism Based on these pharmacological properties domperidone holds research potential in the investigation and management of gastroparesis and as a comparative compound for mechanistic studies of gastric motility disorders