Learn More
Abcam Domperidone (DMSO Solution), 5MG
Catalog No. 502665694
Encompass_Preferred
MW 425.9 Da. Peripheral D₂-like receptor antagonist. Does not readily cross the blood-brain barrier, and displays gastrokinetic and antiemetic properties.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-266-5694 Supplier Abcam Supplier No. AB1474915MG
MW 425.9 Da. Peripheral D₂-like receptor antagonist. Does not readily cross the blood-brain barrier, and displays gastrokinetic and antiemetic properties.
The product is subject to the following: Abcam Restricted Use Statement