Learn More
Abcam Domperidone, Peripheral D2-like receptor antagonist, 500MG
Catalog No. 502707478
Encompass_Preferred
MW 425.9 Da, Purity >99%. Peripheral D₂-like receptor antagonist. Does not readily cross the blood-brain barrier, and displays gastrokinetic and antiemetic properties.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7478 Supplier Abcam Supplier No. AB120566500MG
MW 425.9 Da, Purity >99%. Peripheral D₂-like receptor antagonist. Does not readily cross the blood-brain barrier, and displays gastrokinetic and antiemetic properties.
The product is subject to the following: Abcam Restricted Use Statement