Learn More
Medchemexpress LLC Domperidone (R33812) | 57808-66-9 | 99.9% | 200 MG
SDP
Supplier:  Medchemexpress LLC HYB0411200MG
Encompass_Preferred
Domperidone is an orally active and selective dopamine-2 receptor antagonist. It acts as an antiemetic and a prokinetic agent through its effects on the chemoreceptor trigger zone and motor function of the stomach and small intestine. Unlike Metoclopramide, Domperidone does not cause adverse neurological symptoms due to minimal penetration through the blood-brain barrier, and it is rapidly absorbed after oral administration with few reported side effects.
  • Orally active and selective dopamine-2 receptor antagonist.
  • Acts as an antiemetic and a prokinetic agent.
  • Minimal penetration through the blood-brain barrier.
  • Rapidly absorbed after oral administration.
  • Few reported side effects.
Catalog No. 50-004-06540