Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Domperidone 98.0+%, TCI America™
SDP

Catalog No. D41251G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D41251G 1 g
D4125200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D41251G Supplier TCI America Supplier No. D41251G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 57808-66-9
Molecular Formula C22H24ClN5O2
Molecular Weight (g/mol) 425.917
MDL Number MFCD00069256
InChI Key FGXWKSZFVQUSTL-UHFFFAOYSA-N
Synonym domperidone, motilium, nauzelin, domperidonum, domperidona, domperidon, domperidonum inn-latin, domperidona inn-spanish, motillium, motinorm
PubChem CID 3151
ChEBI CHEBI:31515
IUPAC Name 6-chloro-3-[1-[3-(2-oxo-3H-benzimidazol-1-yl)propyl]piperidin-4-yl]-1H-benzimidazol-2-one
SMILES C1CN(CCC1N2C3=C(C=C(C=C3)Cl)NC2=O)CCCN4C5=CC=CC=C5NC4=O

Specifications

Melting Point 243°C
Color White-Yellow
Quantity 1 g
Formula Weight 425.92
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Domperidone
RTECSNumber DE2275900
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.