Learn More
StatLab Donatello Tissue Processor Series 3
SDP
Catalog No. 22038093
Change view
Click to view available options
For Use With (Application):
IHC, ISH, Molecular Protocols
Catalog No. For Use With (Application)
22-038-093 IHC, ISH, Molecular Protocols
1 options
Catalog No. 22-038-093 Supplier StatLab Supplier No. SDSDT9000
Add to Cart
Add to Cart
An Automatic Tissue Processor that Is Advanced, Reliable and Innovative
  • Preserves cellular detail for clearer results
  • Increases capacity by up to 40% compared to other processors
  • Supports IHC, ISH, and molecular protocols
  • Protects samples during power loss
  • Speeds biopsy processing
  • Ensures consistent safety and quality
  • Backed by expert installation, remote support, and Italian craftsmanship
Specifications
Product Type Tissue Processor
For Use With (Application) IHC, ISH, Molecular Protocols