Learn More
MSC Donegan Optical Headband Magnifier, 1.75x Magnification, 14 in Focal Distance
Catalog No. 5000513450
Encompass
This headband magnifier is a precision binocular optical tool designed to enhance user efficiency and reduce eye strain. It allows for hands-free operation and provides three-dimensional vision, making it suitable for professionals and hobbyists. It can be comfortably worn over prescription or safety eyeglasses.
  • Features optical glass prismatic lenses, ground and polished to precision standards.
  • Lenses are bevel edged and mounted in an interchangeable plastic frame.
  • Adjustable pivots allow the visor to be tilted out of the way when not in use.
  • Headband is adjustable to fit all head sizes and is made from a special blend lightweight material.
  • High impact visor ensures durability and maintains a new appearance.
  • Genuine leather comfort band with padded orthopedic felt provides enhanced comfort.
Catalog No. 50-005-13450 Supplier MSC Supplier No. 00322669
This headband magnifier is a precision binocular optical tool designed to enhance user efficiency and reduce eye strain. It allows for hands-free operation and provides three-dimensional vision, making it suitable for professionals and hobbyists. It can be comfortably worn over prescription or safety eyeglasses.
  • Features optical glass prismatic lenses, ground and polished to precision standards.
  • Lenses are bevel edged and mounted in an interchangeable plastic frame.
  • Adjustable pivots allow the visor to be tilted out of the way when not in use.
  • Headband is adjustable to fit all head sizes and is made from a special blend lightweight material.
  • High impact visor ensures durability and maintains a new appearance.
  • Genuine leather comfort band with padded orthopedic felt provides enhanced comfort.