Learn More
MSC Donegan Optical OptiVISOR Lens Plate, 2.5x, 5 Diopter, 8 in Focal Distance
Catalog No. 5000530004
Encompass
The Donegan Optical LP-5 OptiVISOR Lens Plate is a high-quality optical glass lens designed for precision viewing. With 2.5x magnification, a 5 diopter, and an 8 inch focal distance, this interchangeable lens plate brings work close to the user while reducing eye strain and fatigue, making it suitable for various tasks requiring magnification.
  • Precision viewing brings work close to the user.
  • Worn on the head, allowing both hands to be free for tasks.
  • Adjustable headband ensures a comfortable fit for various head sizes.
  • Lenses are ground and polished to optical standards for accuracy and clarity.
  • Compatible with interchangeable lens plates, offering different magnification options.
  • Designed to minimize eye strain and fatigue during extended use.
  • Ideal for jewelers, watchmakers, artists, hobbyists, and industrial applications.
Catalog No. 50-005-30004 Supplier MSC Supplier No. 00322693
The Donegan Optical LP-5 OptiVISOR Lens Plate is a high-quality optical glass lens designed for precision viewing. With 2.5x magnification, a 5 diopter, and an 8 inch focal distance, this interchangeable lens plate brings work close to the user while reducing eye strain and fatigue, making it suitable for various tasks requiring magnification.
  • Precision viewing brings work close to the user.
  • Worn on the head, allowing both hands to be free for tasks.
  • Adjustable headband ensures a comfortable fit for various head sizes.
  • Lenses are ground and polished to optical standards for accuracy and clarity.
  • Compatible with interchangeable lens plates, offering different magnification options.
  • Designed to minimize eye strain and fatigue during extended use.
  • Ideal for jewelers, watchmakers, artists, hobbyists, and industrial applications.