Learn More
Medchemexpress LLC HY-14566 10mg Medchemexpress, Donepezil CAS:120014-06-4 Purity:>98%
SDP
Catalog No. 502081614
Encompass_Preferred
Medchemexpress, HY-14566 10mg Donepezil CAS:120014-06-4 Donepezil (E2020 free base) is a specific and potent AChE inhibitor with IC50s of 8.12 nM and 11.6 nM for bovine AChE and human AChE, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1614 Supplier Medchemexpress LLC Supplier No. HY1456610MG
Add to Cart
Add to Cart
Medchemexpress, HY-14566 10mg Donepezil CAS:120014-06-4 Donepezil (E2020 free base) is a specific and potent AChE inhibitor with IC50s of 8.12 nM and 11.6 nM for bovine AChE and human AChE, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.