Learn More
Selleck Chemical LLC Donepezil S5073-25mg
SDP
Catalog No. 503108507
Encompass_Preferred
Donepezil (Aricept Donepezilo) is a piperidine based potent specific non-competitive and reversible inhibitor of acetylcholinesterase (AChE) used for the treatment of mild to moderate dementia of the Alzheimer s type
Catalog No. 50-310-8507 Supplier Selleck Chemical LLC Supplier No. S507325MG
Donepezil (Aricept Donepezilo) is a piperidine based potent specific non-competitive and reversible inhibitor of acetylcholinesterase (AChE) used for the treatment of mild to moderate dementia of the Alzheimer s type