Learn More
Medchemexpress LLC Donepezil-d4 hydrochloride | 1219798-88-5 | 100.0% | C24H26D4ClNO3 | 5 MG
SDP
Catalog No. 5000432978
Encompass_Preferred
Donepezil-d4 hydrochloride is a deuterium-labeled form of Donepezil hydrochloride. Donepezil Hydrochloride (E2020) is a reversible, selective AChE inhibitor with an IC50 of 6.7 nM for AChE activity. It demonstrates high selectivity for AChE over BuChE and exhibits a neuroprotective effect on Aβ42 neurotoxicity.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-32978 Supplier Medchemexpress LLC Supplier No. HYB0034S15MG
Donepezil-d4 hydrochloride is a deuterium-labeled form of Donepezil hydrochloride. Donepezil Hydrochloride (E2020) is a reversible, selective AChE inhibitor with an IC50 of 6.7 nM for AChE activity. It demonstrates high selectivity for AChE over BuChE and exhibits a neuroprotective effect on Aβ42 neurotoxicity.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS