Learn More
Medchemexpress LLC Donepezil-d4 hydrochloride (E2020-d4) | 1219798-88-5 | 99.96% | C24H26D4ClNO3 | 1 MG
SDP
Catalog No. 5000433568
Encompass_Preferred
Donepezil-d4 (hydrochloride) is the deuterium labeled Donepezil hydrochloride. Donepezil Hydrochloride (E2020) is a reversible, selective AChE inhibitor with an IC50 of 6.7 nM for AChE activity. Donepezil shows high selectivity for AChE over BuChE and exhibits neuroprotective effect on Aβ42 neurotoxicity.
  • Acts as a reversible, selective AChE inhibitor.
  • Demonstrates high selectivity for AChE over BuChE.
  • Shows neuroprotective effects against Aβ42 neurotoxicity.
  • Can be utilized as a tracer in research.
  • Serves as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes can influence pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-33568 Supplier Medchemexpress LLC Supplier No. HYB0034S11MG
Donepezil-d4 (hydrochloride) is the deuterium labeled Donepezil hydrochloride. Donepezil Hydrochloride (E2020) is a reversible, selective AChE inhibitor with an IC50 of 6.7 nM for AChE activity. Donepezil shows high selectivity for AChE over BuChE and exhibits neuroprotective effect on Aβ42 neurotoxicity.
  • Acts as a reversible, selective AChE inhibitor.
  • Demonstrates high selectivity for AChE over BuChE.
  • Shows neuroprotective effects against Aβ42 neurotoxicity.
  • Can be utilized as a tracer in research.
  • Serves as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes can influence pharmacokinetic and metabolic profiles of drugs.