Learn More
Medchemexpress LLC Donepezil-d7 hydrochloride | 1261394-20-0 | 420782 | 99.0% | C24H23D7ClNO3 | 2.5 MG
SDP
Catalog No. 5000355455
Encompass_Preferred
Donepezil-d7 (hydrochloride) is the deuterium labeled Donepezil. Donepezil (E2020 free base) is a specific and potent AChE inhibitor with IC50s of 8.12 nM and 11.6 nM for bovine AChE and human AChE, respectively. It is for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-55455 Supplier Medchemexpress LLC Supplier No. HY14566S2.5MG
Donepezil-d7 (hydrochloride) is the deuterium labeled Donepezil. Donepezil (E2020 free base) is a specific and potent AChE inhibitor with IC50s of 8.12 nM and 11.6 nM for bovine AChE and human AChE, respectively. It is for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS