Learn More
Selleck Chemical LLC Donepezil HCl S2462-200mg
SDP
Catalog No. 503108831
Encompass_Preferred
Donepezil HCl is a specific and potent AChE inhibitor for bAChE and hAChE with IC50 of 8 12 nM and 11 6 nM respectively This product is not soluble in PBS solution Please do not dissolve it in PBS for administration
Catalog No. 50-310-8831 Supplier Selleck Chemical LLC Supplier No. S2462200MG
Donepezil HCl is a specific and potent AChE inhibitor for bAChE and hAChE with IC50 of 8 12 nM and 11 6 nM respectively This product is not soluble in PBS solution Please do not dissolve it in PBS for administration