Learn More
Medchemexpress LLC Donepezil Hydrochloride | 120011-70-3 | 99.8% | C24H30ClNO3 | 1 G
SDP
Catalog No. 5000432576
Encompass_Preferred
Donepezil Hydrochloride (E2020) is a reversible, selective Acetylcholinesterase (AChE) inhibitor. It demonstrates high selectivity for AChE over Butyrylcholinesterase (BuChE) and exhibits neuroprotective effects against Aβ42 neurotoxicity. Its IC50 for AChE activity is 6.7 nM.
  • Reversible, selective AChE inhibitor
  • High selectivity for AChE over BuChE
  • Exhibits neuroprotective effects against Aβ42 neurotoxicity
Catalog No. 50-004-32576 Supplier Medchemexpress LLC Supplier No. HYB00341G
Donepezil Hydrochloride (E2020) is a reversible, selective Acetylcholinesterase (AChE) inhibitor. It demonstrates high selectivity for AChE over Butyrylcholinesterase (BuChE) and exhibits neuroprotective effects against Aβ42 neurotoxicity. Its IC50 for AChE activity is 6.7 nM.
  • Reversible, selective AChE inhibitor
  • High selectivity for AChE over BuChE
  • Exhibits neuroprotective effects against Aβ42 neurotoxicity