Learn More
TARGETMOL CHEMICALS INC DONEPEZIL HYDROCHLORIDE 200MG
SDP
Catalog No. 502576508
Encompass_Preferred
Also available in 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Donepezil HCl(Aricept) is a selective and effective AChE inhibitor for bAChE and hAChE (IC50 8.12/11.6 nM). purity: 99%
Catalog No. 50-257-6508 Supplier TARGETMOL CHEMICALS INC Supplier No. T6478200MG
Also available in 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Donepezil HCl(Aricept) is a selective and effective AChE inhibitor for bAChE and hAChE (IC50 8.12/11.6 nM). purity: 99%