Learn More
Abcam Donepezil hydrochloride, Acetylcholinesterase inhibitor, 10MG
Catalog No. 502707297
Encompass_Preferred
MW 415.96 g/mol, Purity >99%. Potent, selective acetylcholinesterase inhibitor (IC₅₀ = 5.7 nM). Displays >1250 times selectivity for AChE over butyrylcholinesterase. Centrally active. Neuroprotective activity in vitro and in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7297 Supplier Abcam Supplier No. AB12076310MG
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
MW 415.96 g/mol, Purity >99%. Potent, selective acetylcholinesterase inhibitor (IC₅₀ = 5.7 nM). Displays >1250 times selectivity for AChE over butyrylcholinesterase. Centrally active. Neuroprotective activity in vitro and in vivo.
The product is subject to the following: Abcam Restricted Use Statement