Learn More
Medchemexpress LLC Donitriptan | 170912-52-4 | MFCD09832566 | 98.1% | 403.48 g/mol | C23H25N5O2 | 5 MG
SDP
Catalog No. 5000244001
Encompass_Preferred
Donitriptan is a research compound and a potent, high-efficacy agonist at 5-HT1B and 5-HT1D serotonin receptors, with reported pKi values of about 9.4 and 9.3. It is supplied as a purified small molecule for biochemical and pharmacological studies, with molecular formula C23H25N5O2, molecular weight 403.48 g/mol, and typical purity of approximately 98%.
  • Potent 5-HT1B and 5-HT1D receptor agonist with high affinity.
  • High purity suitable for in vitro pharmacology and biochemical assays.
  • Well characterized molecular properties for reproducible experimental use.
  • Available as the free base form appropriate for standard formulation and testing.
  • Commonly used in serotonergic receptor and migraine-related research applications.
Catalog No. 50-002-44001 Supplier Medchemexpress LLC Supplier No. HY1061575MG
Donitriptan is a research compound and a potent, high-efficacy agonist at 5-HT1B and 5-HT1D serotonin receptors, with reported pKi values of about 9.4 and 9.3. It is supplied as a purified small molecule for biochemical and pharmacological studies, with molecular formula C23H25N5O2, molecular weight 403.48 g/mol, and typical purity of approximately 98%.
  • Potent 5-HT1B and 5-HT1D receptor agonist with high affinity.
  • High purity suitable for in vitro pharmacology and biochemical assays.
  • Well characterized molecular properties for reproducible experimental use.
  • Available as the free base form appropriate for standard formulation and testing.
  • Commonly used in serotonergic receptor and migraine-related research applications.